PCR-microbuizen strips, 0,2 ml met vastgehechte dopstrip, Amplitube™
Leverancier: Simport Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
T322-1B T322-1G T322-1N T322-1R T322-1Y
211-3332EA 153 EUR
211-3332 211-3333 211-3331 211-3334 211-3335
PCR-microbuizen strips, 0,2 ml met vastgehechte dopstrip, Amplitube™
Buisjes PCR-buisjes
PP. Ultra-thin wall design ensures uniform and efficient thermal transfer, therefore reducing PCR reaction time in most V-bottomed thermal cyclers.
  • Tube strips with attached 8-cap strips moulded with a living hinge to facilitate opening and closing
  • Individual tube sealing ensures that samples are well protected from carry-over contamination
  • Choice of natural and four non-cytotoxic and non-metallic colours for visual coding of samples
  • Domed cap offers a snap shut seal to avoid evaporation during thermal cycling
  • Certified RNase, DNase and DNA-free