Reageerbuizen, Durham
Leverancier: MURRAY S
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
G010/22 G010/21 G010/23 G010/24 G011/21 G011/22 G011/23 G011/24
212-0633EA 113 EUR
212-0633 212-7095 212-7096 MURRG010/24 MURRG011/21 212-0448 MURRG011/23 212-0449
Reageerbuizen, Durham
Buisjes Reageerbuisjes
Soda-lime glass test tubes are suitable for general mixing and simple laboratory work, and will withstand moderate warming (water bath). They should not be placed directly into naked flames.
Neutral glass test tubes will withstand moderate temperature change, they should not be taken from cold and placed directly into the hottest part of the flame without pre-heating.
Certificaten: ISO 4142:2002