Reageerbuizen zonder rand, dunwandig
Leverancier: MURRAY S
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
G030/01 G030/02 G030/03 G030/04 G030/05 G030/06 G030/08 G030/10 G030/09
212-7097EA 63.6 EUR
212-7097 212-7098 212-7099 212-7100 212-7102 212-7103 212-0461 212-7104 MURRG030/09
Reageerbuizen zonder rand, dunwandig
Buisjes Reageerbuisjes
Borosilicate test tubes are suitable for all usual laboratory applications. They will withstand temperature change up to and including boiling of samples. They are also highly chemically resistant.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes