Reageerbuizen zonder rand, dikwandig
Leverancier: MURRAY S
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
G033/01 G033/02 G033/03 G033/05 G033/08 G033/10 G033/06
212-7039EA 78.5 EUR
212-7039 212-7040 212-7041 212-7042 212-7043 212-7044 212-0463
Reageerbuizen zonder rand, dikwandig
Buisjes Reageerbuisjes
Borosilicate glass, grade 3.3, SAMCO '33'. Thick walled.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Suitable for all usual laboratory applications. They will withstand temperature change up to and including boiling of samples.
Certificaten: ISO 4142:2002