Reageerbuizen met rand
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1624/10M 1624/12M 1624/16M 1624/20M
212-7421EA 160 EUR
212-7421 212-7422 212-7423 212-7424
Reageerbuizen met rand
Buisjes Reageerbuisjes
PYREX®, borosilicate glass, heavy wall.
  • Excellent chemical resistance
  • High temperature resistance
Heavy wall to provide extra mechanical strength. Robust construction reduces possibility of breakages. Improved user safety.
Certificaten: ISO 4142