Reageerbuisjes met ronde of vlakke bodem, Corning® Gosselin™
Leverancier: Corning
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
TP10-01 TP10-02 TR95-03 TR95-22 TR95-23
212-5164EA 402 EUR
212-5164 212-0680 212-0177 212-0179 212-0180
Reageerbuisjes met ronde of vlakke bodem, Corning® Gosselin™
Buisjes Reageerbuisjes
Transparant PS of doorschijnend PP, met of zonder doppen.
  • With or without cap, aseptic or sterile (10 kGray)
  • Can withstand centrifugation of 3000×g, unless otherwise indicated
  • Suitable for the majority of automated testing systems
Suitable for sampling, transport and centrifugation. Wide variety of diameters and sizes covering all analytical needs.