Reageerbuizen zonder rand
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1626/20M S/3L6016 1626/10M 1626/12M
212-7431EA 464 EUR
212-7431 212-7454 212-7428 212-7429
Reageerbuizen zonder rand
Buisjes Reageerbuisjes
Pyrex®, borosilicate glass, heavy wall.
  • Excellent chemical resistance
  • High temperature resistance
High quality test tubes designed and constructed for optimum performance. Rimless test tubes with a heavy wall to provide extra mechanical strength. Improved user safety.
Certificaten: ISO 4142