Test and sample tubes
Leverancier: Anicrin
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
T275S100 T610S1P0 T610S2P0 T610P1P0
ANICT275S100EA 209 EUR
ANICT275S100 ANICT610S1P0 ANICT610S2P0 ANICT610P1P0
Test and sample tubes
Buisjes Reageerbuisjes
Clear PS or natural-coloured PP.
  • PS tubes: Non-autoclavable. Temperature resistant from −40 to +90 °C
  • PP tubes: Autoclavable up to 122 °C. Temperature resistant from −90 to +135 °C
  • Available with round or conical base
Precision moulded test tubes and sample tubes with round or conical base. Non sterile.
Order Now
Specificaties
Environmentally Preferable