Reageerbuizen met SVL schroefdop
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1636/36MP 1636/10MP 1636/44MP 1636/24MP 1636/26MP 1636/30MP 1636/32MP 1636/34MP 1636/38MP 1636/42MP 1636/04MP
212-7442EA 318 EUR
212-7442 212-7437 212-0655 212-7441 212-7549 212-7550 212-7551 212-7552 212-7553 212-7554 212-7436
Reageerbuizen met SVL schroefdop
Buisjes Reageerbuisjes
PYREX®, borosilicaatglas, fenolische schroefdoppen.
  • Excellent chemical and thermal resistance
  • Screw caps with inert PTFE lined rubber seal
  • Cap can withstand repeated dry heat and steam sterilisation cycles (121 °C for 2 hours)
Reageerbuizen met hals en SVL®-schroefdop.