Reageer- en testbuizen met schroefdop
Leverancier: KIMA SAS DI CHIARIN RENZO
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
17044 17060 17046 17062
212-6713EA 566 EUR
212-6713 212-6719 KIMA17046 KIMA17062
Reageer- en testbuizen met schroefdop
Buisjes Reageerbuisjes
Polystyrene, clear, with red screw cap.
  • Sterile, individually or in the rack with 108 tubes in sterile packed available
  • Non graduated
  • Good thermal and chemical resistance
Universal tubes with round bottom for a wide range of applications.