Reinigingsmiddel voor reageerbuizen
Leverancier: DELTALAB
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
400500 400800 400400 400705 400700 400900 401100 401700
212-3320EA 137 EUR
212-3320 212-3321 212-3322 212-3326 DELT400700 DELT400900 DELT401100 DELT401700
Reinigingsmiddel voor reageerbuizen
Buisjes Reageerbuisjes
Polypropylene (PP), ultra clear.
  • Temperature resistant from 0 to 120 °C (short-term up to 130 °C)
  • Centrifugeable up to 15000×g
  • Autoclavable
Precisely shaped reagent and sample tubes for a variety of applications. With round or flat bottom. Unsterile. Graduated.