Replacement caps
Leverancier: QVF LABORTECHNIK
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
708-68 708-69 708-71
215-2043EA 0 EUR
215-2043 215-2044 215-2046
Replacement caps
Sluitingen Reageerbuissluitingen
Pyrex®, phenoplast, black.
  • Excellent chemical resistance, excellent temperature resistance
  • Screw caps with PTFE-coated seal
  • Replacement seals are available as an option
Replacement closures for reagent and sample glasses as well as bottles with SVL threads.