Test tube holders, wood
Leverancier: GLASWARENFABRIK KARL HECHT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
42811010 42810010
HECH42811010EA 1.9 EUR
HECH42811010 212-6506
Test tube holders, wood
Tangen
Wooden, with spring.
  • Peg/clip shape
Suitable for tubes up to 20/30 mm diameter.