Droogbuizen met NS hals, gehoekt, Quickfit®
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MF23/1 MF23/2 MF23/3 MF23/4
201-0217EA 43.3 EUR
201-0217 201-0218 201-0526 BIBBMF23/4
Droogbuizen met NS hals, gehoekt, Quickfit®
Buisjes Droogbuizen
PYREX®, borosilicate glass with ground glass cone.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Designed for use with a desiccant such as calcium chloride.