Verbinding, kelgel met steel, Quickfit®
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MF15/1 MF15/2 MF15/3
201-3056EA 27.4 EUR
201-3056 201-3057 201-3058
Verbinding, kelgel met steel, Quickfit®
Buisjes Gasinlaatbuizen
Pyrex® borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Cone with stem and reduced shank
Useful for construction of gas inlet tubes.
Stem length: 150 mm
Three shank lengths: 45/60/70 mm.