Buizen, Lucht en stoom toevoer, 90° gebogen, Quickfit®
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MF15/3B/SC MF15/2B/SC
201-8607EA 223 EUR
201-8607 201-3054
Buizen, Lucht en stoom toevoer, 90° gebogen, Quickfit®
Buisjes Gasinlaatbuizen
Pyrex®, borosilicaatglas.
  • Excellent chemical resistance
  • High temperature resistance
Detachable plastic screwthread connector to facilitate the attachment of 9 mm bore flexible tubing. Lower end of tube left plain for glass blower to cut to length and draw to jet.
Stem length: 150 mm