Laboratorium- en pompslangen, Tygon® F-4040-A
Leverancier: Saint Gobain Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
070533-05 070533-08 070533-09 070533-14 070533-15 070533-17 070533-18 070543-12 070543-16 070543-08
224-2334EA 129 EUR
224-2334 224-2338 224-2340 224-2349 224-2350 224-2352 224-2353 224-2315 224-2321 224-2308
Laboratorium- en pompslangen, Tygon® F-4040-A
Slangen Transfer Tubing
Tygon® slangen, geel, doorschijnend.
  • Scheurbestendig, zwellen niet op en worden niet broos
  • Bestand tegen ozon en UV
  • Compatibel met de meeste producten op petroleumbasis
Petroleum and lubricant resistant tubing, that is ideal for the transfer of fuel, kerosene, heating oil, cutting fluids and glycol-based coolants. Does not swell up and harden even when hydrocarbon based liquids are used. This minimises the risk of defects such as cracking or leakage. Due to the low extraction level, tubing also protects the fluids from contamination. Length 381 mm. Distance between stops: 140 mm (2 stops) and 170 mm (3 stops).
Temperature resistance: −35 to +74 °C
Hardheid (Shore A Durometer): 57
Dichtheid: 1,26