Buizen
Leverancier: EMMANUEL PIERRE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1TAR910 1TAR1010 1TAR410 1TAR510 1TAR610 1TAR710 1TAR810
212-0205EA 30.3 EUR
212-0205 212-0199 212-0200 212-0201 212-0202 212-0203 212-0204
Buizen
Slangen Transfer Tubing
Uit AR-glas.
  • Good chemical and physical properties
  • Suitable for the production of items that must normally withstand only brief exposure to aggressive chemicals
  • Not resistant to high thermal loads
Lengte: 500 mm.