Glasbuizen
Leverancier: FISCHER LABORTECHNIK
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
2011003 2011004 2011005 2011006 2011007 2011008 2011009 2011010
201-1003EA 0 EUR
201-1003 201-1004 201-1005 201-1006 201-1007 201-1008 201-1009 201-1010
Glasbuizen
Slangen Transfer Tubing
DURAN®, borosilicate glass.
  • Appropriately cut to 500 mm in length
  • Excellent chemical and thermal resistance
  • Permissible wall thickness tolerance of ±0,2 mm