Non-return valves, SP Bel-Art
Leverancier: Bel-Art Products, a Part of SP
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
H197150000 H197160000 H197170000
229-0538EA 155 EUR
229-0538 BELAH197160000 BELAH197170000
Non-return valves, SP Bel-Art
Ventielen Terugslagventielen
PP valve, translucent with neoprene valve disk. Stops reverse flow of liquid when pressure is interrupted.
  • Useful when connecting a vacuum line to a filter flask
  • Recommended maximum pressure 20 psi. (1034 Torr)
  • Autoclavable