Screw caps with liners, WHEATON®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
242212 242216 242218 242220 242226
548-2040EA 86 EUR
548-2040 548-2041 548-2042 548-2043 548-2044
Screw caps with liners, WHEATON®
Sluitingen Sluitingen voor chromatografievials
These PP screw caps have multi-purpose liners, which are made of three-ply co-extruded foamed low density PE core between two solid layers of low density PE.
  • For use with neutral pH to mild acid and alkali solutions