Aluminium doppen, WHEATON®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
224211-01 224211-06 224211-07 224223-01 224231-01 224235-01 224222-01
218-2102EA 296 EUR
218-2102 WHEA224211-06 WHEA224211-07 218-2109 WHEA224231-01 WHEA224235-01 WHEA224222-01
Aluminium doppen, WHEATON®
Sluitingen Sluitingen voor chromatografievials
Open top seal provides for filling or retrieving contents with a syringe, large target diameter.
  • For use with 12×32 mm crimp top vials
  • Choice of liner
They are also ideal for use as closures in chromatography and instrumentation applications.