Aluminium doppen, WHEATON®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
224192-05 224192-06 224192-07 224193-01 224193-05 224193-06 224193-07 224187-01 224176-01 224183-01 224182-01 224178-05 224178-06 224178-07 224182-05 224182-06 224182-07 224183-05 224183-06 224183-07 224178-01 224191_U 224177-01 224208 224189 224192-01 224197-01 224177-05 W224207 W224205 224191
WHEA224192-05EA 182 EUR
WHEA224192-05 WHEA224192-06 WHEA224192-07 218-2108 WHEA224193-05 WHEA224193-06 WHEA224193-07 218-2118 218-2121 218-2117 218-2114 WHEA224178-05 WHEA224178-06 WHEA224178-07 WHEA224182-05 WHEA224182-06 WHEA224182-07 WHEA224183-05 WHEA224183-06 WHEA224183-07 WHEA224178-01 WHEA224191_U WHEA224177-01 WHEA224208 WHEA224189 WHEA224192-01 218-2134 WHEA224177-05 WHEAW224207 WHEAW224205 WHEA224191
Aluminium doppen, WHEATON®
Sluitingen Sluitingen voor chromatografievials
Aluminum seals (unlined) are available in a wide range of styles and sizes.
  • Open top seals expose stopper for sample retrieval with a syringe
  • Tear-off seal removes completely allowing content to pour from bottle
  • Solid top seals are excellent for long-term storage of samples
  • Flip cap seals provide tamper evidence
Colour selection includes natural, red, blue, and green.