Vials MICRO+™, met glazen inserts
Leverancier: CHROMACOL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
03-FIV 03-FIVA
548-1108EA 0 EUR
548-1108 548-1109
Vials MICRO+™, met glazen inserts
flesjes
All Micro+™ vials have a 300 or 900 µl tapered glass insert fused top and bottom into a standard 12×32 mm vial, which means that analysts no longer have to assemble a number of small components before they can start to work. The insert is now an integral part of the vial, thus reducing the chance of the needle being damaged.