Flesjes voor staalname, WHEATON®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
223687 WUBB2MLC-T-EP WUBB5MLA-T-EP
548-2014EA 269 EUR
548-2014 WHEAWUBB2MLC-T-EP WHEAWUBB5MLA-T-EP
Flesjes voor staalname, WHEATON®
flesjes
These clear glass vials are manufactured from Wheaton '200' low extractable borosilicate glass to maintain uniform wall thickness. The amber vials are manufactured from Wheaton '320'  borosilicate glass.
  • Blowback feature in the neck and a specially designed bottom radius for added strength
  • These features are important for freeze-drying applications
  • Vials are provided in shrink-wrapped modules which greatly reduces particulate contamination
Certificaten: Meets ASTM Type 1, Class A/B and USP Type 1 standards.
Leveringsinformatie: Vials are supplied without closures.
Order Now
Specificaties
Environmentally Preferable