Sluitingen, PPCO, amberkleurig, met kleurcode, 11 mm
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
362825-0112 342825-0110 342825-0111 342825-0112 342825-0113 342825-0114 342825-0115 342825-0116 342825-0117 342825-0118 342825-0119 342825-1110 342825-1111 362825-0111 362825-0113 362825-0114 362825-0115 362825-0116 362825-0117 362825-0118 362825-0119 362825-1110 362825-1111
215-1268EA 309 EUR
215-1268 215-1552 215-1553 215-1560 215-1561 215-1563 215-1564 215-1567 215-1568 215-1569 215-1570 215-1571 215-1583 215-1668 215-1669 215-1670 215-1671 215-1672 215-1679 215-1694 215-1700 215-1701 215-1702
Sluitingen, PPCO, amberkleurig, met kleurcode, 11 mm
Sluitingen Sluitingen voor chromatografievials
Identify samples quickly with amber high profile closures with colour coders for micro packaging vials. Polypropylene polymer closures are ready to use and have no O-ring to fall out or contaminate contents.
  • RNase/DNase free (only sterile closures), non-cytotoxic and non-pyrogenic
  • Permanently attached, colour coded inserts are free of toxic heavy metals
  • Inserts do not come in contact with vial contents
Certificaten: Manufactured in compliance with ISO 13485:2003 quality requirements. Micro packaging vials and closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.
Verpakking: Closures are bulk packed without vials, 500 per inner bag, 1000 per case.