Sluitingen, PPCO, laag profiel, 11 mm
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
362821-0112 342821-0110 342821-1111 342821-0112 342821-0114 342821-0115 342821-0116 342821-0118 342821-0111 362821-0111 362821-1111 362821-0114 362821-0116 362821-0110 362821-0115 362821-0118 362821-1112
215-1265EA 120 EUR
215-1265 215-1192 215-1216 215-1210 215-1211 215-1212 215-1213 215-1215 215-1193 215-1264 215-1267 215-1266 215-1653 215-1651 215-1652 215-1654 NALG362821-1112
Sluitingen, PPCO, laag profiel, 11 mm
Sluitingen Sluitingen voor chromatografievials
Identify samples quickly with low profile closures for micro packaging vials. Polypropylene polymer closures are ready to use.
  • RNase/DNase free (only sterile closures), non-cytotoxic and non-pyrogenic
Certificaten: Manufactured in compliance with ISO 13485:2003 quality requirements. Micro packaging vials and closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.
Verpakking: Closures are bulk packed without vials, 500 per inner bag, 1000 per case.