Sluitingen, PPCO, transparant, met kleurcode, 11 mm
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
342820-0119 342820-0112 342820-0116 342820-0114 362820-0114 342820-0110 342820-0111 342820-0113 342820-0115 342820-0117 342820-0118 342820-1110 362820-0110 362820-0111 362820-0112 362820-0113 362820-0115 362820-0116 362820-0117 362820-0118 362820-0119 362820-1110
215-1191EA 306 EUR
215-1191 215-1188 215-1190 215-1189 215-1263 215-1526 215-1527 215-1528 215-1529 215-1530 215-1531 215-1544 215-1630 215-1631 215-1632 215-1633 215-1638 215-1639 215-1640 215-1648 215-1649 215-1650
Sluitingen, PPCO, transparant, met kleurcode, 11 mm
Sluitingen Sluitingen voor chromatografievials
Identify samples quickly with high profile closures with colour coders for micro packaging vials. Polypropylene polymer closures are ready to use and have no O-ring to fall out or contaminate contents.
  • RNase/DNase free (only sterile closures)
  • Permanently attached, colour coded inserts are free of toxic heavy metals
  • Inserts do not come in contact with vial contents
Certificaten: Manufactured in compliance with ISO 13485:2003 quality requirements. Micro packaging vials and closures meet the requirements of 21CFR177.1520 for food and beverage use, USPMore Product Information