WHEATON® Scintillation Vials
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
986546 986540 986562 986548 986568 986581 986586 986590 986560 986561 986580 986591 986582 986542 986541
218-2249EA 597 EUR
218-2249 548-2170 WHEA986562 215-3909 215-3911 215-3915 215-3927 215-3491 WHEA986560 WHEA986561 215-3913 215-3928 215-3919 218-0000 215-3905
WHEATON® Scintillation Vials
flesjes
These vials are made from WHEATON® 180 low potassium borosilicate glass. The background counts of these vials are consistently low, the ultraviolet transmission is high.
  • Available with attached or unattached screw caps
  • Utility trays can be used to store samples
Certificaten: Glass conforms to ASTM E438 Type I, Class A.
Verpakking: Packed in five utility trays with each partition tray holding 100 vials.