Diagnostic vials, Nalgene®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
2035-0010 2035-0020 2035-0005
391-0075EA 252 EUR
391-0075 391-0076 391-0074
Diagnostic vials, Nalgene®
flesjes
Deze doorzichtige PETG-flacons met witte HDPE-sluitingen zijn ideaal voor bemonstering, opslag en verzending van reagentia of bufferoplossingen.
  • Non-pyrogenic and non-cytotoxic
  • Sterile to 10⁻⁶ SAL
  • 20-415 lined white high-density polyethylene closure attached to PETG vial
  • Leakproof when used with Nalgene closures
  • Narrow mouth
These bottles are constructed from non-pyrogenic and non-cytotoxic polyethylene terephthalate copolymer with lined white HDPE (high-density polyethylene) closures.
Inner neck diameter: 11 mm
Certificaten: Manufactured inMore Product Information
Order Now
Specificaties
Environmentally Preferable Lage impact op de fabricage
Milieu certificering ISO14001 (Certificate Registration No. EMS 752603)
NAR Certificate of Retirement