Horlogeglazen
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1060/04D 1060/06D 1060/18D 1060/20D 1060/14D 1060/21D 1060/09D 1060/10D
216-2770EA 14.8 EUR
216-2770 216-2748 216-2752 216-2777 216-2751 216-2753 BIBB1060/09D BIBB1060/10D
Horlogeglazen
Horlogeglazen
PYREX®, borosilicate glass, with fused edges.
  • Zeer goede chemische bestendigheid
  • Bestand tegen hoge temperaturen
For evaporation work and covering heated beakers. Fire-polished edges to reduce chipping and breakage.