Weegpapier, klasse 2122, Whatman™
Leverancier: Whatman products (Cytiva)
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
10347890 10347893
515-8214EA 140 EUR
515-8214 515-8212
Weegpapier, klasse 2122, Whatman™
Weegpapieren
These Grade 2122 weigh paper sheets reliably allow samples to be transferred to scales without adding unwanted substances that could impact analytical results.
  • Smooth surface allows samples to slide off with ease and not be retained on paper
  • Shiny pergamyne paper allows visual determination that all sample has been removed
  • Extremely thin (0,03 mm) and lightweight (40 g/m²) for safe and secure sample transport
  • No glue or additives (residue-free)
This paper is not low in nitrogen. Therefore, it should not be used for Kjeldahl analysis.