Handdoeken, KLEENEX® en SCOTT®
Leverancier: KIMBERLY CLARK
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
6789 6772 6778 6710
115-1757EA 104 EUR
115-1757 115-1260 115-1935 KIMB6710
Handdoeken, KLEENEX® en SCOTT®
Schoonmaakdoekjes Doekjes en handdoeken voor eenmalig gebruik Papieren handdoekjes
Disposable hand towels made from AIRFLEX*, a voluminous, textile-like fabric, 2 or 3 ply.
  • High absorption performance, and tear proof even when wet
  • Single, self-presenting towels for enhanced hygiene and less waste
  • Extremely soft
Colour: White
KLEENEX® folded towels for dispenser type 6945, 6956 or 8971.
KLEENEX® roll towels for dispenser type 6959 or 6963.