Poetsdoekjes voor laboratium, KIMTECH SCIENCE*
Leverancier: KIMBERLY CLARK
Ratings:
Avg. Ratings: 5.0 (1 Review)
Total Ratings: 1
Avg. Ratings: 5.0 out of 5
Was this review helpful? Yes No
Yes: 6 No: 5 
Laboratoire / gestionnaire de stock / ILE DE FRANCE
Yes, I recommend
Le conditionnement correspond exactement à notre attente, produit très pratique pour notre utilisation.
7551 7552 7552_CH 7551_CH 7557 7558
115-2029EA 386 EUR
115-2029 115-2221 115-2093 KIMB7551_CH 115-2074 115-2075
Poetsdoekjes voor laboratium, KIMTECH SCIENCE*
Schoonmaakdoekjes Doekjes en handdoeken voor eenmalig gebruik Doekjes voor delicate taken
These high quality wipes, made from 100% pure fibre, are suitable for a variety of precision wiping tasks such as the cleaning of delicate glassware, equipment and surfaces. They are also suitable for cleaning liquids and dust, and are equipped with anti-static dispensing to reduce lint and anti-static discharge.
  • Light-duty wipes
  • Low lint absorbency for delicate applications and surfaces
  • Chemically inert and anti-static
  • Supplied in a hygienic, user friendly pop-up box for clean and fast dispensing, even while on the move
Ideal for applications in healthcare, manufacturing, optics and electronics. Suitable for use in ISO Class 4 or higher cleanroom environments.
Colour: White