Poetsdoekjes, Tork Heavy-Duty
Leverancier: Essity
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
530137 530237
115-0275EA 94.7 EUR
115-0275 115-1660
Poetsdoekjes, Tork Heavy-Duty
Schoonmaakdoekjes Herbruikbare reinigingsdoeken Doeken voor algemeen gebruik
Cellulose/PP/polyester, reusable. These low-linting and textured cloths have been specially designed for heavy-duty cleaning and degreasing tasks.
  • Exceptionally durable, thick and bulky
  • Able to absorb and evenly distribute large quantities of solvent
  • Certified for contact with food
The wipes are reusable and need only half the detergents and degreasers required by fabric cloths. For fast, efficient work.
Colours: White or blue
Verpakking: Supplied as rolls, in Top paks or in Handy Boxes.