Poetsdoekjes, Multizell®
Leverancier: Zellstoff-Vertriebs-GmbH & Co. KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
10440B 10440A 10333B 10333A
115-2761EA 83.4 EUR
115-2761 115-2762 115-2763 115-2764
Poetsdoekjes, Multizell®
Schoonmaakdoekjes Herbruikbare reinigingsdoeken Doeken voor algemeen gebruik
These wipes are made from 100% recycled material (cellulose). Ideal to wipe off liquids or to remove soiling. Wipes can be taken one at a time, available in 3 or 4 ply versions and in various sizes.
  • Highly absorbent
  • Lint-free
  • Not bleached