PH-combinatie-elektroden
Leverancier: WTW ONLINE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
109100 109115 109125 109117 109119 109103 109118 109102
662-0280EA 340 EUR
662-0280 708-0036 708-0037 708-0057 708-0058 WTWO109103 WTWO109118 WTWO109102
PH-combinatie-elektroden
Elektroden pH-elektroden
The SensoLyt® measuring chains with arming are easy to install and boast flexible measuring and application areas. Eletrodes without arming can be connected directly to a pH monitor of the EcoLine® or QuadroLine® series. Depending on the application, they can be installed into a flow vessel.
  • Ideal for measurements in water and drinking water
  • Also for high pressure and temperature conditions
  • Easy to maintain by easy measuring chain replacement