pH/mV/°C meters, bench, 765 / 766
Leverancier: KNICK
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
766 766SETB 766SETA 765 765-SET
662-3210EA 1130 EUR
662-3210 662-3211 662-3212 662-3220 662-3221
pH/mV/°C meters, bench, 765 / 766
Elektrochemische meters pH- en multiparametersystemen
Easy-to-use pH-meters with comprehensive safety functions for standard applications in everyday lab routines. Well-designed enclosure with IP 54 protection, waterproof membrane keyboard, drain grooves, and double insulation for electrical safety.
  • Automatic checking of the device functions during power-on
  • Sensoface® feature: electrode condition checking
  • Calimatic® feature: automatic pH calibration (automatic recognition of the pH buffer)
  • Manual or automatic temperature compensation
  • The 765 model also include a standard RS232 interface for a printer