Ontgeurschijven voor autoclaven, Odo-Clave®
Leverancier: Bel-Art Products, a Part of SP
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
H131980002 H131980000 H131980001 H131980003
129-4937EA 285 EUR
129-4937 129-4935 129-4936 129-4938
Ontgeurschijven voor autoclaven, Odo-Clave®
Deodorants en luchtverfrissers
These pads are impregnated with a deodorant that is released by heat, which doesn't merely mask autoclave odours but actually neutralises them.
  • Protective cover melts in autoclave, allowing deodorant to be heat-activated
  • Easy to handle: simply unwrap and place a pad in each bag prior to autoclaving
  • Available in four pleasant scents
B×L: 32×32 mm
Verpakking: 10 schijven per zak, 10 zakken per doos.