Telkamers, Thoma
Leverancier: LABOR OPTIK
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1506000 1510000 1520000 1530000
631-1151EA 62.1 EUR
631-1151 631-1117 631-1118 631-1119
Telkamers, Thoma
Object- en dekglaasjes Telkamers
For counting thrombocytes and erythrocytes, the grid system corresponds to the large square in the middle of the Neubauer chamber.
  • Subdivision doubles the analysis options
  • Bright-line division increases contrast of the net system to make work much easier
  • Spring clips guarantee the exact positioning of the cover slips
  • For blood capillaries
  • Each small square has a surface of 0,0025 mm²