Rotors for Centrifuge 5702/5702 R/5702 RH
Leverancier: EPPENDORF
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
5702720.003 5702704.008 5702700.002 5702705.004 5702740.004 5702746.002 5702747.009
521-0032EA 1160 EUR
521-0032 521-0024 521-0021 521-0025 521-0133 521-0595 521-0597
Rotors for Centrifuge 5702/5702 R/5702 RH
Centrifuge Rotoren
Eppendorf rotors for centrifuge 5702 series - precision in every spin. Tailor your centrifugation with varied capacities and speeds. Accessorise with adaptable buckets for versatile sample processing. Ideal for low-speed applications, ensuring efficiency and reproducibility.
  • Precision-engineered for optimal sample separation
  • All fixed-angle rotors, rotor lids, buckets, caps and adapters can be autoclaved (121 °C, 20 minutes)
  • Accessories (adaptors and buckets) further customise your centrifuge setup
Elevate your low-speed centrifugation with Eppendorf rotors designed for centrifuge 5702 series. These rotors provide a range of capacities and speeds, offering flexibility for various applications. Accessorise with adaptable buckets, ensuring versatility in sample processing. Ideal for low-speed applications, the Centrifuge 5702 series, known for efficiency and reproducibility, is enhanced with these precision-engineered rotors. Explore the range to optimise your centrifuge for reliable performance.