Rotoren voor KR4i / KR25i-centrifuges
Leverancier: JOUAN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
11177397 11177398 11178073 11178111 11178119 11178252 11178255 11177394 11177395 11177392 11178250 11178254 11178119
521-0611EA 7360 EUR
521-0611 521-0612 JOUA11178073 521-0383 521-0384 521-0390 JOUA11178255 521-0608 521-0609 521-0606 JOUA11178250 521-0391 HERA11178119
Rotoren voor KR4i / KR25i-centrifuges
Centrifuge Rotoren
Utilize the rotating capabilities of the Thermo Scientific™ RP6 Windshielded Swinging-Bucket Rotor, a high-capacity, low-speed rotor.
Capacity: 6×1000 ml (6 L) with Round bucket, 12×250 ml (3 L) with Rectangular bucket, Max. speed/RCF: 4250 rpm/5,735×g with Round bucket, 5,392×g with Rectangular rotor.
Order Now
Specificaties
Environmentally Preferable