Manual cell counters
Leverancier: MOINEAU
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1492MW 1494MW 1495/2MW
720-0163EA 144 EUR
720-0163 MOIN1494MW MOIN1495/2MW
Manual cell counters
Celtellers Handmatige celtellers
This manual cell counter can be used in combination of up to five counters.
  • Four digit display (4,3×3,5 mm) with reset knob
  • Use finger pressure
  • Can be fixed in position
W×D×H: 37,6×0,4×38,1 mm