Toebehoren voor behrotest® SMA
Leverancier: BEHR
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
B00218411 B00218412
705-0031EA 6430 EUR
705-0031 705-0032
Toebehoren voor behrotest® SMA
Destructiesystemen Mineralisatiewerkstations
The behrotest® SMA-ARB workstation is a basic system for simultaneous aqua regia digestion of either 6 or 12 samples for heavy metals analysis.
The details of the components of the workstation are described in the table below: