Dispensers, Acurex™ 501, Socorex
Leverancier: Socorex Isba
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
501.02025 501.021 501.022 501.0505 501.051 501.052 501.101 501.102 501.302
613-4837EA 439 EUR
613-4837 613-4838 613-4839 613-4840 613-4841 613-4842 613-4843 613-4844 613-4845
Dispensers, Acurex™ 501, Socorex
Flesdispensers
Variabel volume, autoclaveerbaar.
  • Dosing mechanism protected inside reservoir
  • Retractable graduated column reducing instrument height
  • Fluid path made of inert materials excluding any metal
  • Amber glass adequate for routine use with optimal light protection
  • Choice of four reservoir sizes: 250, 500, 1000 and 2000 ml
Low profile dispensers for safe reagent handling adapted to refrigerator storage as well as water bath heating.
Volume: 0,2 to 30 ml
Leveringsinformatie: Delivered with a QC certificate (lot identification on the instruments).