Gietplaten Mulicaster, Embitec
Leverancier: EMBITEC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
EP-1019 EP-1016 EP-1017 EP-1018 EP-1025
700-4209EA 414 EUR
700-4209 700-4210 700-4211 700-4212 730-0360
Gietplaten Mulicaster, Embitec
Blotkits
Moulded PC. Compatible with the RunOne™ electrophoresis system.
  • Simple to use - no gaskets, clamps or tape required
  • Flexible with options for well size, well configuration, number of wells and resolving area
Order Now
Specificaties
Environmentally Preferable
Gelafmetingen 115×119 115×89