Elektroporatieplaten
Leverancier: BTX
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
45-0466 45-0467 45-0450 45-0452
732-0480EA 133 EUR
732-0480 732-0481 732-0476 732-0477
Elektroporatieplaten
Elektroporatieplaten
Sterile multi-well electroporation plates for transfection/transformation of bacteria, yeast, insect, plant and mammalian cells.