Micro-objectglaasjes
Leverancier: BTX
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
45-0103 45-0104 45-0105 45-0106
732-0004EA 2500 EUR
732-0004 732-0005 732-0006 732-0007
Micro-objectglaasjes
Elektroden Elektroden voor elektroporatie
Microslides have been used for embryo manipulation and cell or protoplast fusion. The Microslide fits on the microscope allowing observation of the alignment and fusion process.
The Microslides are composed of a glass slide and either two strips of stainless steel tubing, which function as electrodes to provide a divergent field, or stainless steel square bars that are set apart at various gaps to provide homogeneous fields.
Compatibiliteit van generator: ECM 830 en ECM 2001.
Field type: Divergent (Microslide with 0,5 or 1,0 mm gap) or homogeneous (Microslide with 3,2 or 10,0 mm gap).
Requires square post cables and coaxial banana plug cable, please contact your local Avantor office for further details.