Verwarmingsplaten, digitaal
Leverancier: NICKEL ELECTRO
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HP1-1D HP1-2D
453-0005EA 2240 EUR
453-0005 453-0007
Verwarmingsplaten, digitaal
Verwarmers Verwarmingsplaten
Digital hotplates with a cast aluminium plate. Precision electronic controller provides increased sensitivity. Stainless steel casing, stove enamelled finish with fitting for optional retort rod.
  • Allow user selectable time and temperature parameters with memory
  • Membrane touch control panel and bright display
  • Fitted with non-slip rubber feet for safety
Timer settings: 0 to 9999 min.
Temperature uniformity: ±0,5 °C at 60 °C.