Analoge hygrometer
Leverancier: Amarell
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Z641760 Z641750
620-1164EA 39.8 EUR
620-1164 AMARZ641750
Analoge hygrometer
Hygrometers Analoge hygrometers
This precision wall hygrometer is suitable for indoor applications. Brass and stainless steel case. Mercury filling.
  • Betrouwbaar model
  • Nauwkeurigheid ±5,0 %